Lithium iodide hydrate, 99%, extra pure
Lithium iodide hydrate, 99%, extra pure
Lithium iodide hydrate, 99%, extra pure
Thermo Scientific Chemicals

Lithium iodide hydrate, 99%, extra pure

CAS: 85017-80-7 | ILi | 133.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 208011000
Price (EUR)
76,50
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
76,50
Each
Chemical Identifiers
CAS85017-80-7
IUPAC Namelithium(1+) iodide
Molecular FormulaILi
InChI KeyHSZCZNFXUDYRKD-UHFFFAOYSA-M
SMILES[Li+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Heavy metals (as Pb)=<15 ppm
Titration Argentometric>=98.5 % (on dry substance)
Sulfate (SO4)=<1000 ppm
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only