Bismuth(III) oxide, 99.9%, (trace metal basis)
Bismuth(III) oxide, 99.9%, (trace metal basis)
Bismuth(III) oxide, 99.9%, (trace metal basis)
Thermo Scientific Chemicals

Bismuth(III) oxide, 99.9%, (trace metal basis)

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 208075000
Price (USD)
237.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
237.00
Each
Chemical Identifiers
CAS1304-76-3
IUPAC Namedibismuth(3+) trioxidandiide
Molecular FormulaBi2O3
InChI KeyTYIXMATWDRGMPF-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Bi+3].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric88.4 to 91.0 % (Bi)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only