L(+)-Histidine monohydrochloride monohydrate, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L(+)-Histidine monohydrochloride monohydrate, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets JP, BP and EP specifications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2081-061 kg
2081-0712 kg
2081-0950 kg
Catalog number 2081-06
Price (KRW)
-
Quantity:
1 kg
This product is GMP Manufactured and meets JP, BP and EP specifications.
This product is GMP Manufactured and meets JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS5934-29-2
Chemical Name or MaterialL(+)-Histidine
ColorColorless
Melting Point259°C
Molecular FormulaC6H9N3O2·HCl·H2O
PackagingPoly bottle, tamper-evident cap
SynonymH-His-OH.HCl.H2O, L-Histidine monohydrochloride monohydrate, (S)-(+)-Histidine monohydrochloride monohydrate, L-Histidine hydrochloride hydrate, His HCl monohydrate
GradeMulti-Compendial
Molecular Weight (g/mol)209.63
Percent Purity98.5 to 101%
Quantity1 kg
pH3 to 5
Unit SizeEach