L(+)-Amethopterin hydrate, 99%
L(+)-Amethopterin hydrate, 99%
L(+)-Amethopterin hydrate, 99%
Thermo Scientific Chemicals

L(+)-Amethopterin hydrate, 99%

L(+)-Amethopterin hydrate, CAS # 133073-73-1, is an effective inhibitor of dihydrofolate reductase, which may have anti-tumor activity. | CAS: 133073-73-1 | C20H22N8O5 | 454.45 g/mol
数量:
25 mg
100 mg
500 mg
パッケージ:
Glass Bottle
製品番号(カタログ番号) 208100250
価格(JPY)
-
数量:
25 mg
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS133073-73-1
IUPAC Name(2S)-2-[(4-{[(2,4-diaminopteridin-6-yl)methyl](methyl)amino}phenyl)formamido]pentanedioic acid
Molecular FormulaC20H22N8O5
InChI KeyFBOZXECLQNJBKD-UGPWUYPHNA-N
SMILESCN(CC1=CN=C2N=C(N)N=C(N)C2=N1)C1=CC=C(C=C1)C(=O)N[C@@H](CCC(O)=O)C(O)=O
さらに表示
Appearance (Color)Yellow to orange
HPLC>=98.5 %
Specific optical rotation+19° to +24° (20°C, 589 nm) (c=1, 14 g/l Na2CO3)
Water=<13 % (K.F.)
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L(+)-Amethopterin hydrate, is a potent inhibitor of dihydrofolate reductase
  • It has tumoricidal activity

Applications

  • L(+)-Amethopterin hydrate is an inhibitor of dihydrofolate reductase
  • It can be used in anti-tumor studies
  • It is a cell cycle arrest agent, which can inhibit synthesis of DNA, RNA, and proteins
  • It can also inhibit production of pro-inflammatory cytokines
RUO – Research Use Only

General References:

  1. Friedman, B.; Cronstein, B. Methotrexate mechanism in treatment of rheumatoid arthritis. Joint Bone Spine. 2019, 86(3), 301-307.
  2. Sasso, S.P.; Gilli, R.M.; Sari, J.C.; Rimet, O.S.; Briand, C.M. Thermodynamic study of dihydrofolate reductase inhibitor selectivity. Biochim Biophys Acta. 1994, 20, 1207(1), 74-9.
  3. Pittman, S.M.; Strickland, D.; Ireland, C.M. Polymerization of tubulin in apoptotic cells is not cell cycle dependent. Exp Cell Res. 1994, 215 (2), 263-72.