1-Chloro-4-fluorobenzene, 98%
1-Chloro-4-fluorobenzene, 98%
1-Chloro-4-fluorobenzene, 98%
Thermo Scientific Chemicals

1-Chloro-4-fluorobenzene, 98%

CAS: 352-33-0 | C6H4ClF | 130.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 208200250
Price (USD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS352-33-0
IUPAC Name1-chloro-4-fluorobenzene
Molecular FormulaC6H4ClF
InChI KeyRJCGZNCCVKIBHO-UHFFFAOYSA-N
SMILESFC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Refractive index1.4940 to 1.4960 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only