Thermo Scientific™
Eight-Stage Non-Viable Andersen Cascade Impactor
 Related applications:
Environmental
Sample wet or dry particulates with the Thermo Scientific™ Eight-Stage Non-Viable Andersen Cascade Impactor. First in its class, the impactor is the original design cited as the world's reference for aerosol particle sizing. The impactors are uniquely defined as a primary standard classification device thanks to performance verification through well-established fundamental physical principles. It is a multi-stage, multi-orifice sampler used in environmental working areas to measure the size distribution and mass concentration levels of solid particulates and liquid aerosols.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
The combination of a constant flow rate and successively smaller diameter orifices increase the velocity of sample air as it cascades through the sampler, resulting in the impaction of progressively smaller particles in the succeeding stages. Operating at 28.3 liters per minute (LPM), the particle fraction ranges from 0.4 to >10.0 mirometers in diameter. Particles too small to be impacted on the last collection plate are collected in the backup filter. Optional accessories allow for additional research operations at 60 and 90 PLM.
Key Features:
  • Gravimetric analysis allows reference method precision
  • Ease of operation and calibration
  • Particle bounce and wall losses virtually eliminated
  • High mass collection and high flow rate
  • Gravimetric or chemical sample analysis
  • Stainless-steel, glass, and filter substrates available
Additional Information:
  • Each sampler comprises eight aluminum stages held together by three spring clamps and gasketed with O-ring seals. Each contains multiple precision-drilled orifices.
  • Ambient gases enter the inlet cone and cascade through the succeeding orifice stages with successively higher orifice velocities from Stage 0 to Stage 7.
  • Successively smaller particles are inertially impacted onto the collection plates. Submicrometer particles, which are not collected by the last collection plate, are caught in the backup filter, which is an integral part of the impactor immediately downstream from Stage 7.
  • Stage 0 is an orifice stage only, while Stage 8 is a collection stage only. Each sampler requires a flow rate of exactly 28.3L/min. (1 cfm) for maximum efficiency.
Recommended for:
  • Pharmaceutical applications
  • Studying ventilation
  • Indoor or outdoor air quality
  • Hazardous waste sites
  • Emergency response situations
User services
Order status 
Review your order status.
Service offerings
Overview of all services including support plan options, repairs and calibrations, spare parts and technical training.
Factory Depot 
Request an RMA number and send your instrument to our factory for repair.
Talk Tech Q&A Newsletter
Receive answers from the experts
Our dedicated, customer-focused technical support team solves problems with customers each and every day and we are pleased to now bring this information directly to you through Talk Tech, Q&A ambient particulate solutions.
Documents
Manuals & protocols