Zirconyl chloride octahydrate, 98%
Zirconyl chloride octahydrate, 98%
Zirconyl chloride octahydrate, 98%
Thermo Scientific Chemicals

Zirconyl chloride octahydrate, 98%

CAS: 13520-92-8 | CH16Cl2O9 | 243.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 208371000
Price (EUR)
108,65
Online exclusive
120,00
Save 11,35 (9%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
108,65
Online exclusive
120,00
Save 11,35 (9%)
Each
Chemical Identifiers
CAS13520-92-8
Molecular FormulaCl2H16O9Zr
InChI KeySDFOCNSOSARAKC-UHFFFAOYSA-L
SMILESO.Cl[Zr](Cl)=O
Molecular Weight (g/mol)322.24
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric97.5 to 102.5 %
Solubility(10% in water) Clear to slightly hazy solution
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Water42 to 50 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only