L(+)-Lysine monohydrochloride, 98.5-101.5%, powder, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L(+)-Lysine monohydrochloride, 98.5-101.5%, powder, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2084-0950 kg
2084-061 kg
Catalog number 2084-09
Price (MXN)
-
Quantity:
50 kg
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS657-27-2
Chemical Name or MaterialL(+)-Lysine monohydrochloride
ColorWhite/Beige
Melting Point263°C to 264°C
Molecular FormulaH2N(CH2)4CH(NH2)CO2H·HCl
PackagingPolyethylene drum, open head
Synonym(S)-(+)-2,6-Diaminohexanoic acid monohydrochloride, H-Lys-OH.HCl, L-Lysine monohydrochloride, L-Lysine monohydrocholoride, (S)-(+)-Lysine monohydrochloride
GradeMulti-Compendial/USP
Molecular Weight (g/mol)182.65
Percent Purity98.5 to 101.5%
Quantity50 kg
Unit SizeEach