L-Methionine, 98.5-101.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L-Methionine, 98.5-101.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
2085-05100 g
2085-061 kg
Número de catálogo 2085-05
Precio (MXN)
-
Cantidad:
100 g
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Punto de ebullición278°C (1013 hPa)
CAS63-68-3
Nombre del producto químico o materialL-Methionine
ColorWhite
Punto de fusión136°C
Fórmula molecularC5H11NO2S
EnvasePoly bottle, tamper-evident cap
Sinónimo(S)-(-)-2-Amino-4-(methylthio)butyric acid, L(-)-Methionine, Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH
GradoMulticomponente/USP
Peso molecular (g/mol)149.21
Porcentaje de pureza98.5 to 101.5%
Cantidad100 g
pH5.6 to 6.1
Unit Size100 g