L-Methionine, 98.5-101.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L-Methionine, 98.5-101.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2085-061 kg
2085-05100 g
Catalog number 2085-06
Price (MXN)
-
Quantity:
1 kg
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Specifications
Boiling Point278°C (1013 hPa)
CAS63-68-3
Chemical Name or MaterialL-Methionine
ColorWhite
Melting Point136°C
Molecular FormulaC5H11NO2S
PackagingPoly bottle, tamper-evident cap
Synonym(S)-(-)-2-Amino-4-(methylthio)butyric acid, L(-)-Methionine, Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH
GradeMulti-Compendial/USP
Molecular Weight (g/mol)149.21
Percent Purity98.5 to 101.5%
Quantity1 kg
pH5.6 to 6.1
Unit SizeEach