L(-)-Proline, 98.5-101.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L(-)-Proline, 98.5-101.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Have Questions?
Catalog NumberQuantity
2087-061 kg
Catalog number 2087-06
Price (MXN)
-
Quantity:
1 kg
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Specifications
Boiling Point245°C
CAS147-85-3
Chemical Name or MaterialL(-)-Proline
ColorWhite
Melting Point220°C to 222°C
Molecular FormulaC5H9NO2
PackagingPoly bottle, tamper-evident cap
Synonym(S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
GradeMulti-Compendial/USP
Molecular Weight (g/mol)115.13
Percent Purity98.5 to 101.5%
Quantity1 kg
Unit SizeEach