Methyl orotate, 95%
Methyl orotate, 95%
Methyl orotate, 95%
Thermo Scientific Chemicals

Methyl orotate, 95%

CAS: 6153-44-2 | C6H6N2O4 | 170.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 208780250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6153-44-2
IUPAC Namemethyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Molecular FormulaC6H6N2O4
InChI KeyUUTDWTOZAWFKFW-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(=O)NC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Appearance (Color)White to yellow to light brown
Titration with NaOH94.0 to 106.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only