Ammonium dichromate, 99%
Ammonium dichromate, 99%
Ammonium dichromate, 99%
Thermo Scientific Chemicals

Ammonium dichromate, 99%

CAS: 7789-09-5 | H8Cr2N2O7 | 252.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 208810025
Price (USD)
437.65
Special offer
Online exclusive
Ends: 28-Dec-2026
515.00
Save 77.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
437.65
Special offer
Online exclusive
Ends: 28-Dec-2026
515.00
Save 77.35
Each
Chemical Identifiers
CAS7789-09-5
IUPAC Namediammonium [(oxidodioxochromio)oxy]chromiumoylolate
Molecular FormulaCr2H8N2O7
InChI KeyJOSWYUNQBRPBDN-UHFFFAOYSA-P
SMILES[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Appearance (Form)Adhering crystals
Loss on drying=<3 % (105°C, 4 h)
Chloride (Cl)=<500 ppm
Titration Iodometric>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only