L-Threonine, 98.5-101.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L-Threonine, 98.5-101.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
2089-0712 kg
2089-061 kg
Número de catálogo 2089-07
Precio (MXN)
-
Cantidad:
12 kg
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Especificaciones
CAS72-19-5
Nombre del producto químico o materialL-Threonine
ColorColorless
Punto de fusión255°C (Decomposes)
EnvasePolyethylene pail, screw top
Sinónimo(2S,3R)-(-)-Threonine, (2S,3R)-2-Amino-3-hydroxybutyric acid, H-Thr-OH
GradoMulticomponente/USP
Peso molecular (g/mol)119.12
Porcentaje de pureza98.5 to 101.5%
Cantidad12 kg
pH5 to 6.5
Unit SizeEach