Limited time offer: 3 year warranty when you purchase a SimpliAmp Thermal Cycler. View flyer »
Pierce™ Citraconic Anhydride
Catalog number: 20907B
Thermo Scientific™  Related applications: Protein Labeling & Crosslinking
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Thermo Scientific Pierce Citraconic Anhydride reacts with primary amines to to form an amide bond with a terminal carboxyl group. This modification can be reversed using acidic conditions (pH 3-4) or with hydroxylamine.
Citraconic Anhydride (2-methylmaleic anhydride) is used to block primary amine groups at alkaline pH values (pH 7-9). At acidic pH (3-4), the amide linkage is hydrolyzed and citraconic acid is released, revealing the original amine.
Features of Citraconic Anhydride:
• Temporarily block amines to derivatize other parts of the molecule
• Unblock amines using acidic conditions or hydroxylamine
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Reactivity: Amine
Label or Dye: Unreactive Group
Product Line: Pierce™
Product Size: Custom
Contents & storage
Store in a cool, dry area, protected from direct sunlight.
Documents
Manuals & protocols