3-Nitrobenzenesulfonic acid, sodium salt, 97%
3-Nitrobenzenesulfonic acid, sodium salt, 97%
3-Nitrobenzenesulfonic acid, sodium salt, 97%
Thermo Scientific Chemicals

3-Nitrobenzenesulfonic acid, sodium salt, 97%

CAS: 127-68-4 | C6H4NNaO5S | 225.15 g/mol
数量:
1 kg
2.5 kg
パッケージ:
すべて表示
Plastic drum
製品番号(カタログ番号) 209120025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Plastic drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS127-68-4
IUPAC Namesodium 3-nitrobenzene-1-sulfonate
Molecular FormulaC6H4NNaO5S
InChI KeyLJRGBERXYNQPJI-UHFFFAOYSA-M
SMILES[Na+].[O-][N+](=O)C1=CC=CC(=C1)S([O-])(=O)=O
さらに表示
Appearance (Form)Powder or crystalline powder
Appearance (Color)White to yellow
Infrared spectrumConforms
Water=<3.0 % (K.F.)
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only