L(-)-Tryptophan, 98.5-101.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L(-)-Tryptophan, 98.5-101.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.Más información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
2092-0712 kg
2092-061 kg
Número de catálogo 2092-07
Precio (USD)
-
Cantidad:
12 kg
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
CAS73-22-3
Nombre del producto químico o materialL(-)-Tryptophan
ColorWhite
Punto de fusión290°C
Fórmula molecularC11H12N2O2
EnvasePolyethylene pail, screw top
SinónimoL-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH, L-Tryptophan, Tryptophan
GradoMulticomponente/USP
Peso molecular (g/mol)204.23
Porcentaje de pureza98.5 to 101.5%
Cantidad12 kg
pH5.5 to 7
Unit SizeEach