Vinylmagnesium bromide, 0.7M solution in THF, AcroSeal™
Vinylmagnesium bromide, 0.7M solution in THF, AcroSeal™
Vinylmagnesium bromide, 0.7M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Vinylmagnesium bromide, 0.7M solution in THF, AcroSeal™

CAS: 1826-67-1 | C2H3BrMg | 131.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 209398000
Price (TWD)
-
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS1826-67-1
IUPAC Namebromo(ethenyl)magnesium
Molecular FormulaC2H3BrMg
InChI KeyXHHHAXOHMKAOSL-UHFFFAOYSA-M
SMILESBr[Mg]C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Titration Grignard0.70 to 0.90 M
AppearanceMay develop some turbidity or precipitate
Appearance (Color)Clear yellow to orange to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only