L-Tyrosine disodium salt dihydrate, ≥98%, BAKER ANALYZED™, Biochemical Reagent, GMP, J.T.Baker™
Avantor™

L-Tyrosine disodium salt dihydrate, ≥98%, BAKER ANALYZED™, Biochemical Reagent, GMP, J.T.Baker™

For laboratory, research, or manufacturing use.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
2094-05100 g
2094-0712 kg
Número de catálogo 2094-05
Precio (CLP)
-
Cantidad:
100 g
For laboratory, research, or manufacturing use.
For laboratory, research, or manufacturing use.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
CAS122666-87-9
Nombre del producto químico o materialL-Tyrosine disodium salt dihydrate
ColorWhite
Punto de fusión200°C
Fórmula molecularC9H9NO3Na2·2H2O
EnvasePoly bottle, tamper-evident cap
SinónimoDisodium 2-Amino-3-(4-oxidophenyl) propanoate, L-3-(4-Hydroxyphenyl)alanine disodium salt
GradoBioquímica
Peso molecular (g/mol)261.9
Porcentaje de pureza≥98%
Cantidad100 g
pH10.5 to 11.5 (25°C, 1% aqueous solution)
Unit Size100 g