Specifications
CAS122666-87-9
Chemical Name or MaterialL-Tyrosine disodium salt dihydrate
ColorWhite
Melting Point200°C
Molecular FormulaC9H9NO3Na2·2H2O
PackagingPolyethylene pail, screw top
SynonymDisodium 2-Amino-3-(4-oxidophenyl) propanoate, L-3-(4-Hydroxyphenyl)alanine disodium salt
GradeBiochemical
Molecular Weight (g/mol)261.9
Percent Purity≥98%
Quantity12 kg
pH10.5 to 11.5 (25°C, 1% aqueous solution)
Unit Size12 kg