L(+)-Valine, 98.5-101.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L(+)-Valine, 98.5-101.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2095-05100 g
2095-061 kg
2095-0712 kg
Catalog number 2095-05
Price (MXN)
-
Quantity:
100 g
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point217°C (1013 hPa)
CAS72-18-4
Chemical Name or MaterialL(+)-Valine
ColorWhite
Melting Point86°C
Molecular FormulaC5H11NO2
PackagingPoly bottle, tamper-evident cap
Synonym(S)-(+)-Valine, Valine, H-Val-OH
GradeMulti-Compendial/USP
Molecular Weight (g/mol)117.15
Percent Purity98.5 to 101.5%
Quantity100 g
pH5.5 to 7
Unit SizeEach