1,2-Bis(trimethylsilyloxy)ethane, 98%
1,2-Bis(trimethylsilyloxy)ethane, 98%
1,2-Bis(trimethylsilyloxy)ethane, 98%
Thermo Scientific Chemicals

1,2-Bis(trimethylsilyloxy)ethane, 98%

CAS: 7381-30-8 | C8H22O2Si2 | 206.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 209580100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7381-30-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4020 to 1.4040 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only