Ethylenediaminetetraacetic acid, dipotassium salt dihydrate, 99%, extra pure
Ethylenediaminetetraacetic acid, dipotassium salt dihydrate, 99%, extra pure
Ethylenediaminetetraacetic acid, dipotassium salt dihydrate, 99%, extra pure
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, dipotassium salt dihydrate, 99%, extra pure

CAS: 25102-12-9 | C10H14K2N2O8·2H2O | 404.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 209741000
Price (EUR)
48,65
Online exclusive
53,40
Save 4,75
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
48,65
Online exclusive
53,40
Save 4,75
Each
Chemical Identifiers
CAS25102-12-9
IUPAC Namedipotassium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular FormulaC10H18K2N2O10
InChI KeyJFROQFOCFOKDKU-UHFFFAOYSA-L
SMILESO.O.[K+].[K+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Chloride (Cl)=<50 ppm
Solubility(10 % in water) Clear colorless
Heavy metals (as Pb)=<10 ppm
Iron (Fe)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only