Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, dipotassium salt dihydrate, 99%, extra pure

Catalog number: 209745000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, dipotassium salt dihydrate, 99%, extra pure

Catalog number: 209745000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 209745000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25102-12-9
IUPAC Name
dipotassium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular Formula
C10H18K2N2O10
InChI Key
JFROQFOCFOKDKU-UHFFFAOYSA-L
SMILES
O.O.[K+].[K+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Heavy metals (as Pb)
=<10 ppm
Iron (Fe)
=<10 ppm
Infrared spectrum
Conforms
Chloride (Cl)
=<50 ppm
Solubility
(10 % in water) Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text