Benzaldehyde dimethyl acetal, 98%
Benzaldehyde dimethyl acetal, 98%
Benzaldehyde dimethyl acetal, 98%
Thermo Scientific Chemicals

Benzaldehyde dimethyl acetal, 98%

CAS: 1125-88-8 | C9H12O2 | 152.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 209792500
Price (USD)
111.65
Online exclusive
124.00
Save 12.35
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
111.65
Online exclusive
124.00
Save 12.35
Each
Chemical Identifiers
CAS1125-88-8
IUPAC Name(dimethoxymethyl)benzene
Molecular FormulaC9H12O2
InChI KeyHEVMDQBCAHEHDY-UHFFFAOYSA-N
SMILESCOC(OC)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.4910 to 1.4940 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only