Explosion-Proof Freezer, 5.5 Cu. Ft./155.7L, 230 V/50 Hz, 3 Adjustable Shelves, Bottom Storage
Explosion-Proof Freezer, 5.5 Cu. Ft./155.7L, 230 V/50 Hz, 3 Adjustable Shelves, Bottom Storage
Thermo Scientific™

Explosion-Proof Freezer, 5.5 Cu. Ft./155.7L, 230 V/50 Hz, 3 Adjustable Shelves, Bottom Storage

Maintain a safe work environment with Thermo Scientific™ Explosion-Proof Refrigerators, engineered to prevent triggering an explosion inside or outside the unit.
Have Questions?
Catalog number 20EREETSV
Price (USD)
-

Prevent the triggering of an explosion inside or outside the unit with Thermo Scientific™ Explosion-Proof Refrigerators, suitable for use in Class I, Division 1, Group C and D hazaRDO™us environments. Thermostat and compressor components are isolated within a vapor-proof enclosure, and electrical connections are hardwired to prevent sparking and accidental disconnection.

  • Hydraulic thermostat
  • Manual defrost
  • Magnetic vinyl gaskets for positive door seal
  • CFC-free insulation and refrigerant
  • Warranty varies by country, please contact us for details
Specifications
DescriptionExplosion-Proof Refrigerator
TypeExplosion-Proof Refrigerator
Capacity (English)20 cu. ft.
Capacity (Metric)566.34 L
Temperature Range1°C to 12°C
Temperature (Setpoint)Adjustable
RefrigerantR134a, 100% CFC Free
Door StyleHinged
No. of Doors1 (1 compartment)
Hinge SideRight
Shelves4 Fixed
Door Shelves6 Fixed
Exterior FinishEpoxy Coated Steel
InsulationPolyurethane Foam
InteriorEpoxy Coated Steel
DisplayTemperature Display - None
CastersAdjustable Leveling Feet
ColorWhite
Control TypeMechanical Thermostat/Outside, On Top Of Unit, In Blue Housing
For Use With (Application)Indoor Cold Storage Of Materials In Class I, Division I Group C and D, Class I Zone 1 Group IIB Environments
Typical Lifetime10 Years
WarrantyWarranty varies by country, please contact us for details
DefrostManual
LightingInterior - No
Min. Clearance3 in. On Sides, 3 in. At Back, 3 in. On Top
Model20EREETSV
Access SecurityDoor Lock
CompressorBottom Mounted
Dimensions (D x W x H) Interior23.8 x 27.8 x 57.8 in. (60.3 x 70.5 x 146.7 cm)
Dimensions (L x W x H) Exterior32.13 x 34 x 73.13 in. (81.6 x 86.4 x 186.1 cm)
Amperage2.0 A
Plug TypeNot Provided
Voltage230 V
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Wattage186 w
Shipping Weight (English)283 lb.
Shipping Weight (Metric)128 kg
Unit SizeEach