Search Thermo Fisher Scientific
This product has been discontinued and replaced with the RO20AA and RadEye G20-10.
Certifications/Compliance | IEC 60846 |
Description | Range-Locking Model |
Battery Type | 2 D Alkaline |
Measurement Range | 0 m to 500 mSv h-1 (0 to 50 R/h) |
Unit Size | Each |
Description | Two Integrating Ranges |
Battery Type | 2 D Alkaline |
Chamber Bias | Yes |
Battery Life | Clock back-up using very long-life Lithium cells |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (EUR) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2100SEID | Each | Range-Locking Model | Contact Us › | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2120S | Each | Two Integrating Ranges | Contact Us › | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
This product has been discontinued and replaced with the RO20AA and RadEye G20-10.