Orion™ Online ATC Probe
Actual product may vary
Orion™ Online ATC Probe
Orion™ Online ATC Probe
Orion™ Online ATC Probe
Thermo Scientific™

Orion™ Online ATC Probe

Take pH measurements that are automatically corrected for variation in electrode slope due to temperature change with this online ATC probe.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberResistanceLength (Metric) Cable
2100TP30,000ohms0.6 m
2001TM1000ohms5 m
2001XT30,000ohms10 m
Catalog number 2100TP
Price (USD)
458.10
Online Exclusive
509.00
Save 50.90 (10%)
Each
Resistance:
30,000ohms
Length (Metric) Cable:
0.6 m
Price (USD)
458.10
Online Exclusive
509.00
Save 50.90 (10%)
Each

Take pH measurements that are automatically corrected for variation in electrode slope due to temperature change with the Thermo Scientific™ Orion™ Online ATC Probe. This probe features stripped, nonterminated ends and an unbreakable stainless-steel body to withstand rigorous use.

  • Designed for use with the Orion 2001 and Orion 2002 online pH analzyers
  • Used with single and multiple buffer standardization procedures
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionATC Probe, 30K with .5M Cable
For Use With (Equipment)2100 Platform
Length (English) Cable1.97 ft.
Length (Metric) Cable0.6 m
Resistance30,000ohms
Temperature (Metric) Range0°C to 100°C
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeProbe
pH Range0 to 14
Unit SizeEach