Pierce™ Gentle Ag/Ab Elution Buffer, pH 6.6

Catalog number: 21013B

Thermo Scientific™  Related applications: Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce IgG Binding and Elution Buffers ensure high-yield, nondenaturing antibody purification with IgG affinity purification supports, such as Protein A and Protein G agarose beads. The Gentle Ag/Ab Binding and Elution Buffer system uses near-neutral, high-salt conditions for elution, enabling even sensitive and labile antibodies (or other proteins) to be eluted from an affinity system without denaturation and inactivation. The phosphate-free binding buffer in this system is a necessary substitute for the Protein A binding buffer (pH 8), which precipitates with the Gentle Ag/Ab Elution Buffer.

Shared features of all Thermo Scientific Pierce IgG Binding and Elution Buffers:

Ready to use—carefully formulated and prefiltered 1X solutions
Three binding buffers—optimized for Protein A, Protein G or Protein A/G
Two elution buffer options—acidic (pH 2.8) or high salt (near-neutral)
Versatile—elution buffers are appropriate for antibody-antigen affinity purification

Products available for each antibody purification method:
Protein A IgG Binding Buffer: pH 8.0; phosphate-based; contains glycerol and EDTA
Protein A/G IgG Binding Buffer: pH 8.0; phosphate-based; contains EDTA
Protein G IgG Binding Buffer: pH 5.0; contains 0.02% sodium azide
IgG Elution Buffer: pH 2.8; amine-based
Gentle Ag/Ab Binding Buffer: pH 8.0; phosphate-free; contains 0.05% sodium azide
Gentle Ag/Ab Elution Buffer: pH 6.6; high salt (ionic strength)

Related Products
Pierce™ IgG Elution Buffer
Pierce™ Gentle Ag/Ab Binding Buffer, pH 8.0
Pierce™ Gentle Ag/Ab Binding and Elution Buffer Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Line: Pierce™
Product Size: Custom
Reagent Type: Elution Buffer

Contents & storage

Store at 4°C.

Documents

Manuals & protocols