Nalgene™ Wide-Mouth Lab Quality HDPE Bottles
Nalgene™ Wide-Mouth Lab Quality HDPE Bottles
Nalgene™ Wide-Mouth Lab Quality HDPE Bottles
Thermo Scientific™

Nalgene™ Wide-Mouth Lab Quality HDPE Bottles

Perform countless field applications utilizing these general-purpose, wide-mouth, lab quality HDPE bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)QuantityUnit Size
2104-0008PK
also known as 2104-0008
250 mL12/Pk.Pack of 12
2104-000130 mL12/Pk.Case of 72
2104-0001PK
also known as 2104-0001
30 mL12/Pk.Pack of 12
2104-000260 mL72/Pk.Case of 72
2104-0002PK
also known as 2104-0002
60 mL72/Pk.Pack of 12
2104-0004125 mL12/Pk.Case of 72
2104-0004PK
also known as 2104-0004
125 mL12/Pk.Pack of 12
2104-0008250 mL12/Pk.Case of 72
2104-0016500 mL12/Pk.Case of 48
2104-0016PK
also known as 2104-0016
500 mL12/Pk.Pack of 12
2104-00321 L6/Pk.Case of 24
2104-0032PK
also known as 2104-0032
1 L6/Pk.Pack of 6
2104-00481500 mL24/Pk.Case of 24
2104-0048PK
also known as 2104-0048
1500 mL24/Cs.Pack of 6
Catalog number 2104-0008PK
also known as 2104-0008
Price (EUR)
31,20
Pack of 12
Capacity (Metric):
250 mL
Quantity:
12/Pk.
Unit Size:
Pack of 12
Price (EUR)
31,20
Pack of 12

Utilize Thermo Scientific™ Nalgene™ Wide-Mouth Lab Quality HDPE Bottles for countless field applications. These containers exhibit excellent chemical resistance to most acids, bases and alcohols and are suitable for freezer use to -100°C and are Leakproof† when used with Nalgene closures. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

Features:

  • Excellent chemical resistance to most corrosives make these bottles versatile and indispensable in your lab
  • Ideal for storing, shipping, and packaging liquids — uniform walls provide durability and resistance to splitting or punctures
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
  • Leakproof† when used with Nalgene closures
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials

Includes:

Polypropylene screw closure

Recycling Information:

Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
ACT LabelYes
Capacity8 oz., 250 mL
Capacity (English)8 oz.
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure ColorNatural
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size43 mm
Closure TypeScrew Cap
DescriptionLab Quality Wide-Mouth Bottle
Diameter (Metric)61 mm
Diameter (Metric) Inner Neck33 mm
DisposableReusable
Height (English)5.16 in.
Height (Metric)131 mm
Neck Finish43 mm
No. per Pack12/Pk.
Product TypeWide Mouth Lab Quality Bottle
Quantity12/Pk.
ShapeRound
Through the Closure DispensingNo
Width (English)2.4 in.
Capacity (Metric)250 mL
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialHDPE
MouthWide
SterilityNon-sterile
Unit SizePack of 12

Frequently asked questions (FAQs)

Why are your Nalgene containers and caps always different materials?

In order to function as a leakproof system without a liner (one of the key advantages of our laboratory and packaging products), the surfaces of the container and closure that meet and push together when tightened must be dissimilar materials with different hardnesses. Even our polypropylene copolymer (PPCO) bottles with polypropylene homopolymer (PP) bottles are different enough to function properly. On some larger carboy products, this difference cannot be sufficiently maintained so a pliable thermoplastic elastomer (TPE) gasket is added for proper leakproof seal function.