Silver iodide, 99%, pure
Silver iodide, 99%, pure
Silver iodide, 99%, pure
Thermo Scientific Chemicals

Silver iodide, 99%, pure

CAS: 7783-96-2 | AgI | 234.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 210420050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7783-96-2
IUPAC Namesilver(1+) iodide
Molecular FormulaAgI
InChI KeyMSFPLIAKTHOCQP-UHFFFAOYSA-M
SMILES[Ag+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
Assay>=98.5 % (ex Ag)
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only