Silver(I) fluoride, 99+%
Silver(I) fluoride, 99+%
Silver(I) fluoride, 99+%
Thermo Scientific Chemicals

Silver(I) fluoride, 99+%

CAS: 7775-41-9 | AgF | 126.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Plastic bottle
Catalog number 210450250
Price (USD)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7775-41-9
IUPAC Namesilver(1+) fluoride
Molecular FormulaAgF
InChI KeyREYHXKZHIMGNSE-UHFFFAOYSA-M
SMILES[F-].[Ag+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown-yellow to rust brown
Assay>=99.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only