Ethylmagnesium bromide, 0.9M solution in THF, AcroSeal™
Ethylmagnesium bromide, 0.9M solution in THF, AcroSeal™
Ethylmagnesium bromide, 0.9M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Ethylmagnesium bromide, 0.9M solution in THF, AcroSeal™

CAS: 925-90-6 | C2H5BrMg | 133.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 210478000
Price (EUR)
309,00
Each
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (EUR)
309,00
Each
Chemical Identifiers
CAS925-90-6
IUPAC Namebromo(ethyl)magnesium
Molecular FormulaC2H5BrMg
InChI KeyTWTWFMUQSOFTRN-UHFFFAOYSA-M
SMILESCC[Mg]Br
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Grignard0.85 to 1.15 M
Appearance (Color)Clear to slightly turbid colorless to yellow
Appearance (Form)Solution
Appearance (Color)to brown to grey-brown to brown-black
AppearanceMay develop some turbidity or precipitate
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only