How can I increase detection sensitivity with SuperSignal West Femto Maximum Sensitivity Substrate?
Treating the nitrocellulose or PVDF membrane with Western Blot Signal Enhancer (Cat. No. 21050) before blocking can boost the chemiluminescent signal of low-intensity bands.