L-Glutamine, 100 g - FAQs
View additional product information for L-Glutamine - FAQs (21051024, 21051040)
1 product FAQs found
Why did my 200 mM L-glutamine precipitate when I thawed it?
When L-glutamine is in a concentrated stock solution it easily precipitates when cooled. Warming the solution briefly in a 37 degrees C water bath with gentle swirling will dissolve the precipitate. Do not use the product unless the precipitate is fully dissolved.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.