2,2-Dimethyl-1,3-propanediamine, 99%
2,2-Dimethyl-1,3-propanediamine, 99%
2,2-Dimethyl-1,3-propanediamine, 99%
Thermo Scientific Chemicals

2,2-Dimethyl-1,3-propanediamine, 99%

CAS: 7328-91-8 | C5H14N2 | 102.18 g/mol
数量:
50 g
250 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 210510500
価格(JPY)
-
数量:
50 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7328-91-8
IUPAC Name2,2-dimethylpropane-1,3-diamine
Molecular FormulaC5H14N2
InChI KeyDDHUNHGZUHZNKB-UHFFFAOYSA-N
SMILESCC(C)(CN)CN
さらに表示
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Low melting solid or liquid
Titration with HClO498.5 to 101.5 %
Appearance (Color)Colorless to white to almost white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only