Cyclohexane, 99.5%, extra pure
Cyclohexane, 99.5%, extra pure
Cyclohexane, 99.5%, extra pure
Thermo Scientific Chemicals

Cyclohexane, 99.5%, extra pure

CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Plastic Drum
Catalog number 210570100
Price (EUR)
349,00
Each
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (EUR)
349,00
Each
Chemical Identifiers
CAS110-82-7
IUPAC Namecyclohexane
Molecular FormulaC6H12
InChI KeyXDTMQSROBMDMFD-UHFFFAOYSA-N
SMILESC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.01 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
Residue after evaporation=<0.005 %
Color scale=<10 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only