2,2-Diethoxyacetophenone, 96%
2,2-Diethoxyacetophenone, 96%
2,2-Diethoxyacetophenone, 96%
Thermo Scientific Chemicals

2,2-Diethoxyacetophenone, 96%

CAS: 6175-45-7 | C12H16O3 | 208.26 g/mol
数量:
5 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 210621000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6175-45-7
IUPAC Name2,2-diethoxy-1-phenylethan-1-one
Molecular FormulaC12H16O3
InChI KeyPIZHFBODNLEQBL-UHFFFAOYSA-N
SMILESCCOC(OCC)C(=O)C1=CC=CC=C1
さらに表示
Appearance (Color)Clear colorless to dark yellow
Appearance (Form)Liquid
Refractive index1.4980 to 1.5010 (20°C, 589 nm)
Infrared spectrumConforms
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only