Search
View additional product information for Pierce™ Background Eliminator Kit - FAQs (21065)
2 product FAQs found
We offer the Pierce Background Eliminator Kit (Cat. No. 21065) that reduces high background, shading, speckling, and over-exposed bands on exposed film.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
We offer the Pierce Background Eliminator Kit (Cat. No. 21065) that reduces high background, shading, speckling, and over-exposed bands on exposed film.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.