Difco™ Phytone™ Supplement, ultra-filtered (UF)
Actual product may vary
Gibco™

Difco™ Phytone™ Supplement, ultra-filtered (UF)

Difco Phytone, ultra-filtered (UF), is an animal origin-free enzymatic digest of soy. Due to the naturally high concentrations of vitaminsRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
210931500 g
21093610 kg
Catalog number 210931
Price (USD)
224.00
Each
Quantity:
500 g
Price (USD)
224.00
Each
Difco Phytone, ultra-filtered (UF), is an animal origin-free enzymatic digest of soy. Due to the naturally high concentrations of vitamins and carbohydrates in soy, Difco Phytone UF is a nutritious and excellent carbohydrate source. This product is an ultra-filtered version of Phytone Peptone, making it desirable for use with mammalian cell culture applications where reduced endotoxin levels are important.

Features:
• Ultra-filtered
• Animal origin-free nutritional supplement
• Excellent carbohydrate source due to the naturally high concentrations of vitamins and carbohydrates in soy

Applications of Difco Phytone, ultra-filtered (UF), include:
• Desirable for use with mammalian cell culture applications where reduced endotoxin levels are important
• Recommended for use in media for the cultivation of a wide variety of organisms, including fungi

Specifications
Certifications/ComplianceLUO
Culture TypeMammalian, Insect, Microbial
For Use With (Application)Cell Culture
FormatBottle
Product LineDifco
Quantity500 g
Shelf Life1460 Days
SourceAnimal origin-free
FormDehydrated
Product TypeMedia Supplement
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.