Difco Phytone, ultra-filtered (UF), is an animal origin-free enzymatic digest of soy. Due to the naturally high concentrations of vitaminsRead more
Have Questions?
Change view
Catalog Number
Quantity
210936
10 kg
210931
500 g
2 Options
Catalog number 210936
Price (EUR)
3.405,00
Each
Quantity:
10 kg
Price (EUR)
3.405,00
Each
Difco Phytone, ultra-filtered (UF), is an animal origin-free enzymatic digest of soy. Due to the naturally high concentrations of vitamins and carbohydrates in soy, Difco Phytone UF is a nutritious and excellent carbohydrate source. This product is an ultra-filtered version of Phytone Peptone, making it desirable for use with mammalian cell culture applications where reduced endotoxin levels are important.
Features: • Ultra-filtered • Animal origin-free nutritional supplement • Excellent carbohydrate source due to the naturally high concentrations of vitamins and carbohydrates in soy
Applications of Difco Phytone, ultra-filtered (UF), include: • Desirable for use with mammalian cell culture applications where reduced endotoxin levels are important • Recommended for use in media for the cultivation of a wide variety of organisms, including fungi
Specifications
Culture TypeMammalian, Insect, Microbial
For Use With (Application)Cell Culture
Product LineDifco
Quantity10 kg
SourceAnimal origin-free
Product TypeDehydrated Culture Media
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.