Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Thermo Scientific™

Nalgene™ Square Wide-Mouth PPCO Bottles with Closure

Simplify cleaning, sample filling and retrieval with these space-efficient, wide-mouth PPCO bottles, which are contact-clear, autoclavable and resistant to most acids, alcohols and bases.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeUnit Size
2110-0008250 mL43 mmCase of 72
2110-000260 mL28 mmCase of 72
2110-0002PK
also known as 2110-0002
60 mL28 mmPack of 12
2110-0006175 mL38 mmCase of 72
2110-0006PK
also known as 2110-0006
175 mL38 mmPack of 12
2110-0008PK
also known as 2110-0008
250 mL43 mmPack of 12
2110-0016500 mL53 mmCase of 48
2110-0016PK
also known as 2110-0016
500 mL53 mmPack of 12
2110-00321 L63 mmCase of 24
2110-0032PK
also known as 2110-0032
1 L63 mmPack of 6
Catalog number 2110-0008
Price (KRW)
306,000
Case of 72
Capacity (Metric):
250 mL
Closure Size:
43 mm
Unit Size:
Case of 72
Price (KRW)
306,000
Case of 72

Save valuable lab space with the square design of Thermo Scientific™ Nalgene™ Wide-Mouth Square Polypropylene Copolymer Bottles. Wide-mouth design allows easy cleaning, sample filling and retrieval. Made from only high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.

Features:
  • Resistant to acids, alcohols and bases
  • Contact-clear bottle provides visible liquid level
  • Square design provides a large area for labeling
  • Linerless polypropylene closures provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Autoclavable
  • Leakproof† when used with Nalgene closures
  • Manufactured from non-toxic materials

Note:

Completely disengage threads or remove cap before autoclaving.

.

Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
AutoclavableAutoclavable
Capacity8 oz., 250 mL
Capacity (English)8 oz.
Certifications/ComplianceUSP Class VI; FDA - 21 CFR
Closure ColorNatural, translucent
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size43 mm
Closure TypeScrew Cap
DescriptionWide-Mouth Square Bottle
Diameter (English)2.4 in.
Diameter (English) Inner Neck1.3 in.
Diameter (Metric)61 mm
Diameter (Metric) Inner Neck33 mm
Height (English)4.6 in.
Height (Metric)116 mm
No. per Case72
No. per Pack12
Product TypeSquare Wide Mouth Bottle
ShapeSquare
Through the Closure DispensingNo
Width (English)2.44 in.
Capacity (Metric)250 mL
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialPPCO
MouthWide
SterilityNon-sterile
Unit SizeCase of 72

Frequently asked questions (FAQs)

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.

Why are your Nalgene containers and caps always different materials?

In order to function as a leakproof system without a liner (one of the key advantages of our laboratory and packaging products), the surfaces of the container and closure that meet and push together when tightened must be dissimilar materials with different hardnesses. Even our polypropylene copolymer (PPCO) bottles with polypropylene homopolymer (PP) bottles are different enough to function properly. On some larger carboy products, this difference cannot be sufficiently maintained so a pliable thermoplastic elastomer (TPE) gasket is added for proper leakproof seal function.