Sulfato de hierro(II) heptahidrato, 99 %, para análisis, Thermo Scientific Chemicals
Sulfato de hierro(II) heptahidrato, 99 %, para análisis, Thermo Scientific Chemicals
Sulfato de hierro(II) heptahidrato, 99 %, para análisis, Thermo Scientific Chemicals
Sulfato de hierro(II) heptahidrato, 99 %, para análisis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfato de hierro(II) heptahidrato, 99 %, para análisis, Thermo Scientific Chemicals

CAS: 10101-97-0 | NiO46H2O | 262.85 g/mol
Have Questions?
Número de catálogoCantidadEnvase
21108010010 kgTambor de plástico
Número de catálogo 211080100
Precio (USD)
-
Cantidad:
10 kg
Envase:
Tambor de plástico
Pedido a granel o personalizado
Identificadores químicos
CAS10101-97-0
IUPAC Namenickel(2+) hexahydrate sulfate
Molecular FormulaH12NiO10S
InChI KeyRRIWRJBSCGCBID-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Crystals
Titration Complexometric98.5 to 101.5 %
pH4 to 6 (5 % at 20°C)
Chloride (Cl)=<50 ppm
Arsenic (As)=<10 ppm
Ver más

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only