Consumables and Services Kits for Orion™ 2110XP Ammonia Analyzer
Actual product may vary
Thermo Scientific™

Consumables and Services Kits for Orion™ 2110XP Ammonia Analyzer

Keep needed supplies and service items in stock to avoid lost productivity with Thermo Scientific™ Consumables and Services Kits for Orion™ 2110XP Ammonia Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
2110XPCALAmmonia Calibration Kit
210056Reference Electrode Half-Cell
211050XPAmmonia Consumables Kit
211050XPFReagentless Consumables Kit
2110XPEKAmmonia Electrode Kit
2110XPEPElectronics Faceplate
2110XPSKService Kit
Catalog number 2110XPCAL
Price (INR)
-
Type:
Ammonia Calibration Kit
Keep needed supplies and service items in stock to avoid lost productivity with Thermo Scientific™ Consumables and Services Kits for the Thermo Scientific™ Orion™ 2110XP Ammonia Analyzer.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Accuracy (Temperature)±0.5 °C
Accuracy (mV)±(0.5 mV + 0.1%)
DescriptionAmmonia Calibration Kit with Carrying Case
Display Type2110XPCAL
Flow Rate40 mL/min.
Includes0.5mL pipette (204847-001), box of pipette tips (204846-001), ammonia calibration standards kit (181040), wash bottle and inlet filter
Inlet Pressure8 to 100 psig
Inputs85-132; 170-264 VAC
OutputsAlarm: 3; Relay: 3; Analog: 2; Selections: 0-20mA; 4-20 mA
Range (mV)±1999.9 m
Relay DetailsMax Load: 250VAC; 30VDC; 5A
Resolution (Temperature)0.1 °C
Resolution (mV)0.1 mV
Response Time2110XPCAL
Sample Inlet0.25 NPTF Tube Fitting in.
Sample Temperature5 to 45 °C
Temperature (Metric) Operating-10°C to +120°C
TypeAmmonia Calibration Kit
Unit SizeEach