Thermo Scientific Chemicals

Silver hexafluorophosphate, 98%, -6 mesh, Thermo Scientific Chemicals

Catalog number: 211120250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Silver hexafluorophosphate, 98%, -6 mesh, Thermo Scientific Chemicals

Catalog number: 211120250
25 g, Plastic bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
26042-63-7
IUPAC Name
silver(1+) hexafluoro-λ⁵-phosphanuide
Molecular Formula
AgF6P
InChI Key
SCQBROMTFBBDHF-UHFFFAOYSA-N
SMILES
[Ag+].F[P-](F)(F)(F)(F)F
Silver (Ag)
>=41.6 %
Appearance (Form)
Powder
Appearance (Color)
White to grey-beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text