N-(Benzyloxycarbonyloxy)succinimide, 98%
N-(Benzyloxycarbonyloxy)succinimide, 98%
N-(Benzyloxycarbonyloxy)succinimide, 98%
Thermo Scientific Chemicals

N-(Benzyloxycarbonyloxy)succinimide, 98%

CAS: 13139-17-8 | C12H11NO5 | 249.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 211170250
Price (USD)
59.65
Online exclusive
65.80
Save 6.15
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
59.65
Online exclusive
65.80
Save 6.15
Each
Chemical Identifiers
CAS13139-17-8
IUPAC Namebenzyl 2,5-dioxopyrrolidin-1-yl carbonate
Molecular FormulaC12H11NO5
InChI KeyMJSHDCCLFGOEIK-UHFFFAOYSA-N
SMILESO=C(OCC1=CC=CC=C1)ON1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
HPLC>=97.5 %
Appearance (Form)Crystalline powder
Melting point77°C to 82°C
Loss on drying=<0.25 % (60°C, 2 h, 1 mmHg)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only