(-)-Diisopropyl D-tartrate, 99%
(-)-Diisopropyl D-tartrate, 99%
(-)-Diisopropyl D-tartrate, 99%
Thermo Scientific Chemicals

(-)-Diisopropyl D-tartrate, 99%

CAS: 62961-64-2 | C10H18O6 | 234.25 g/mol
Have Questions?
Catalog NumberQuantityPackaging
21118010010 gGlass bottle
Catalog number 211180100
Price (CLP)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS62961-64-2
IUPAC Name1,4-bis(propan-2-yl) (2R,3S)-2,3-dihydroxybutanedioate
Molecular FormulaC10H18O6
InChI KeyXEBCWEDRGPSHQH-OCAPTIKFSA-N
SMILESCC(C)OC(=O)[C@@H](O)[C@@H](O)C(=O)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4380 to 1.4400 (20°C, 589 nm)
Specific optical rotation-16° to -18.5° (20°C, 589 nm) (neat)
Appearance (Color)Clear colorless
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only