Accessories and Kits for Orion™ 2111LL Low Level Sodium Analyzers
Actual product may vary
Accessories and Kits for Orion™ 2111LL Low Level Sodium Analyzers
Accessories and Kits for Orion™ 2111LL Low Level Sodium Analyzers
Thermo Scientific™

Accessories and Kits for Orion™ 2111LL Low Level Sodium Analyzers

Have all the auxiliary items on hand for top production with these accessories and kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
211192FORReagentless Consumables Kit
181148Sodium Calibration Kit
21003MCable Kit
210048Sodium Half-Cell Electrode
210058Half-Cell Reference Electrode
211192Sodium Consumables Kit
211194Tubing Kit
2111EKElectrode Kit
2111MKMaintenance Kit
Catalog number 211192FOR
Price (USD)
-
Type:
Reagentless Consumables Kit
Have all the auxiliary items on hand for top production. We offer an assortment of accessories and kits for the Thermo Scientific™ Orion™ 2111LL Low Level Sodium Analyzers. Accessories and kits include calibration kit and carrrying case, analyzer enclosure, service kit, maintenance kit, consumable kit, and related electrodes.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Accuracy±5% or 0.01ppb – whichever is greater
DescriptionReagent-less Low Level Sodium Consumables Kit - for one year operation
Flow Rate40 mL/min.
For Use With (Equipment)Orion™ 2111LL Low Level Sodium Analyzers
Frequency50/60 Hz
Includes6 x 0.8L empty bottles for use with diisopropylamine (DIPA) reagent, 6 diffusion tubing kits (211194), reagent bottle adapter, sodium calibration standards kit with etch solution (181148), sodium electrode (210048), reference electrode (210058), 5 x 2 oz. reference filling solution (211172), O-ring kit (2111LLOK), check valve (2111VC), box of pipette tips (207525-001), restriction tube assembly and 2 x 60 micron inlet filters with gaskets (181170)
Inlet Pressure8 to 100 psig
Max. Relative Humidity211192FOR
Outputs211192FOR
Range0.001-10 ppb-ppm
Resolution1; 2; 3; or 4 significant digits
Response Time95% within 2 minutes
Sample Temperature5 to 45 °C
Voltage85/132 V (100 mA), 170/264 V (200 mA)
Weight (English)50 lb.
Weight (Metric)22.7 g
Temperature (Metric) Operating5°C to 45°C
TypeReagentless Consumables Kit
Unit SizeEach