Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Actual product may vary
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Thermo Scientific™

Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers

Adapt your sodium analyzer to specific needs with Thermo Scientific™ Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzer.
Have Questions?
變更視圖buttonViewtableView
產品號碼Type
2111XPCALSodium Calibration Kit
210045Sodium ISE
211150XPCation/High Acid Consumables
211150XPFReagentless Cation/High Acid Kit
211160XPAmmonia Consumables Kit
211160XPFReagentless Ammonia Kit
211192XPDIPA Consumables Kit
211192XPFReagentless DIPA Consumables Kit
2111XAAmmonia Application Package
2111XCCation/High Acid Package
2111XDDIPA Application Package
2111XPEKElectrode Kit
2111XPEPElectronics Faceplate
產品號碼 2111XPCAL
價格 (TWD)
-
Type:
Sodium Calibration Kit
Adapt your sodium analyzer to your specific analytical needs with Thermo Scientific™ Application Packages, Consumables and Replacement Parts for the Thermo Scientific™ Orion™ 2111XP Sodium Analyzer.
Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
規格
Case MaterialValox 264
Certifications/Compliance2111XPCAL
DescriptionSodium Calibration Kit with Carrying Case
Detection Limits0.1 ppb
Display TypeCustom backlit LCD with temperature, concentration, mV, error codes, menu driven prompts
Flow Rate40 mL/min.
For Use With (Equipment)Orion™ 2111XP Sodium Analyzer
Includes0.1mL pipette, box of pipette tips, wash bottle, 2 oz. each low level and high level sodium standards (19.1ppm, 192ppm, 1910ppm and 19200ppm) and 1 oz. sodium electrode etch solution
Inlet Pressure8 to 100 psig
OutputsAnalog: 3; Alarm: 3; Relay: 3
Relay DetailsMax Load: 250VAC; 30VDC; 5A
Response Time95% within 2 minutes
Sample Inlet1/4 in. NPTF tube fitting
Sample Temperature5 to 45 °C
Voltage85/132 VAC, 170/264 VAC
Temperature (Metric) Operating5°C to 45°C (Sample)
TypeSodium Calibration Kit
Unit SizeEach