Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Actual product may vary
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Thermo Scientific™
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Adapt your sodium analyzer to specific needs with Thermo Scientific™ Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
2111XPEKElectrode Kit
210045Sodium ISE
211150XPCation/High Acid Consumables
211150XPFReagentless Cation/High Acid Kit
211160XPAmmonia Consumables Kit
211160XPFReagentless Ammonia Kit
211192XPDIPA Consumables Kit
211192XPFReagentless DIPA Consumables Kit
2111XAAmmonia Application Package
2111XCCation/High Acid Package
2111XDDIPA Application Package
2111XPCALSodium Calibration Kit
2111XPEPElectronics Faceplate
Catalog number 2111XPEK
Price (USD)
1,278.00
Online exclusive
1,420.00
Save 142.00
Each
Type:
Electrode Kit
Price (USD)
1,278.00
Online exclusive
1,420.00
Save 142.00
Each
Adapt your sodium analyzer to your specific analytical needs with Thermo Scientific™ Application Packages, Consumables and Replacement Parts for the Thermo Scientific™ Orion™ 2111XP Sodium Analyzer.
Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description2111LL Electrode Kit
For Use With (Equipment)Orion™ 2111XP Sodium Analyzer
Includes2111LL Electrode Kit - includes sodium electrode, reference electrode and 2 oz. reference electrode filling solution
TypeElectrode Kit
Unit SizeEach